Assay Details
Target Gene Details
Entrez Gene ID: | 10462 |
Gene Name: | C-type lectin domain family 10 member A |
Gene Aliases: |
CD301, CLECSF13, CLECSF14, HML, HML2, MGL |
Location: |
Chr.17:7074537-7080635 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC10A | NM_006344.2 | 9 | 1253 | NP_006335.2 |
| NM_182906.2 | 9 | 1325 | NP_878910.1 | |
| XM_005256412.1 | 9 | 1270 | XP_005256469.1 | |
| XM_011523613.1 | 9 | 1360 | XP_011521915.1 | |
| XM_011523614.2 | 10 | 1426 | XP_011521916.1 | |
| XM_011523615.1 | 9 | 1348 | XP_011521917.1 | |
| BC027858.1 | 7 | 1592 | AAH27858.1 | |
| BC039011.1 | 9 | 1273 | AAH39011.1 | |
| BM974488.1 | 1 | 464 | ||
| BU687211.1 | 1 | 219 | ||
| D50532.1 | 9 | 1157 | BAA09101.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv5167 | Chr.17:7069362 - 7077999 on Build GRCh38 | Loss |
|
| esv2669929 | Chr.17:7047097 - 7091663 on Build GRCh38 | Deletion |
|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map