Assay Details
Target Gene Details
Entrez Gene ID: | 60494 |
Gene Name: | coiled-coil domain containing 81 |
Gene Aliases: |
- |
Location: |
Chr.11:86374736-86423109 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC81 | NM_001156474.1 | NP_001149946.1 | ||
| NM_021827.4 | NP_068599.3 | |||
| XM_005274161.1 | XP_005274218.1 | |||
| XM_005274162.3 | XP_005274219.1 | |||
| XM_017018105.1 | XP_016873594.1 | |||
| AK027167.1 | BAB15681.1 | |||
| AK131331.1 | BAD18491.1 | |||
| AK223168.1 | BAD96888.1 | |||
| AK298875.1 | ||||
| BC126412.1 |

Genomic Map