Assay Details
Target Gene Details
Entrez Gene ID: | 3398 |
Gene Name: | inhibitor of DNA binding 2, HLH protein |
Gene Aliases: |
GIG8, ID2A, ID2H, bHLHb26 |
Location: |
Chr.2:8681983-8684453 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ID2 | NM_002166.4 | NP_002157.2 | ||
| BC030639.2 | AAH30639.1 |
More Information
Set Membership: |
|

Genomic Map