Assay Details
Target Gene Details
Entrez Gene ID: | 51102 |
Gene Name: | mitochondrial trans-2-enoyl-CoA reductase |
Gene Aliases: |
CGI-63, FASN2B, NRBF1 |
Location: |
Chr.1:29192657-29230958 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MECR | NM_001024732.2 | 10 | 1516 | NP_001019903.2 |
| NM_016011.3 | 10 | 1389 | NP_057095.3 | |
| XM_005245885.2 | 11 | 1439 | XP_005245942.1 | |
| XM_005245887.2 | XP_005245944.1 | |||
| XM_011541539.1 | XP_011539841.1 | |||
| XM_011541540.1 | XP_011539842.1 | |||
| XM_011541541.1 | XP_011539843.1 | |||
| XM_011541543.1 | XP_011539845.1 | |||
| XM_011541545.1 | XP_011539847.1 | |||
| XM_011541546.1 | XP_011539848.1 | |||
| XM_011541547.1 | XP_011539849.1 | |||
| XM_011541549.1 | XP_011539851.1 | |||
| XM_011541550.1 | XP_011539852.1 | |||
| XM_011541553.1 | XP_011539855.1 | |||
| XM_011541554.1 | XP_011539856.1 | |||
| XM_017001411.1 | XP_016856900.1 | |||
| XM_017001412.1 | XP_016856901.1 | |||
| XM_017001413.1 | XP_016856902.1 | |||
| XM_017001414.1 | XP_016856903.1 | |||
| XM_017001415.1 | XP_016856904.1 | |||
| XM_017001416.1 | XP_016856905.1 | |||
| AF151821.1 | AAD34058.1 | |||
| AK025830.1 | ||||
| AK095099.1 | 10 | 1482 | ||
| AK225687.1 | 10 | 1355 | ||
| BC001419.2 | AAH01419.1 |
More Information
Set Membership: |
|

Genomic Map