Assay Details
Target Gene Details
Entrez Gene ID: | 51129 |
Gene Name: | angiopoietin like 4 |
Gene Aliases: |
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158 |
Location: |
Chr.19:8364127-8374375 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL4 | NM_001039667.2 | NP_001034756.1 | ||
| NM_139314.2 | NP_647475.1 | |||
| NR_104213.1 | ||||
| XM_005272484.3 | 7 | 1292 | XP_005272541.1 | |
| XM_005272485.3 | 6 | 1178 | XP_005272542.1 | |
| AB056477.1 | BAB40692.1 | |||
| AF153606.1 | AAD41088.1 | |||
| AF169312.1 | AAF62868.1 | |||
| AF202636.1 | AAG22490.1 | |||
| AK222489.1 | BAD96209.1 | |||
| AK222524.1 | BAD96244.1 | |||
| AK303269.1 | ||||
| AY358275.1 | AAQ88642.1 | |||
| BC023647.2 | AAH23647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643614 | Chr.19:8333390 - 8391885 on Build GRCh38 | Gain |
|
| esv3643616 | Chr.19:8366946 - 8383577 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map