Assay Details
Target Gene Details
Entrez Gene ID: | 338879 |
Gene Name: | ribonuclease A family member 10 (inactive) |
Gene Aliases: |
RAH1, RNASE9 |
Location: |
Chr.14:20510472-20511122 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE10 | NM_001012975.1 | 1 | 534 | NP_001012993.1 |
| AK296766.1 | 4 | 1022 | ||
| AY665805.1 | 1 | 534 | AAV87183.1 | |
| BC133002.1 | 1 | 553 | ||
| BC133028.1 | 1 | 553 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map