Assay Details
Target Gene Details
Entrez Gene ID: | 56914 |
Gene Name: | otoraplin |
Gene Aliases: |
FDP, MIAL1 |
Location: |
Chr.20:16748353-16752164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OTOR | NM_020157.3 | 1 | 90 | NP_064542.1 |
| XM_017027959.1 | XP_016883448.1 | |||
| AF233261.1 | 1 | 85 | AAF82078.1 | |
| AF243505.1 | AAG42356.1 | |||
| AJ242552.1 | CAC27443.1 | |||
| AY359082.1 | 1 | 79 | AAQ89441.1 | |
| BC101688.1 | 1 | 90 | AAI01689.1 | |
| BC101690.1 | 1 | 90 | AAI01691.1 |
More Information
Set Membership: |
|

Genomic Map