Assay Details
Target Gene Details
Entrez Gene ID: | 820 |
Gene Name: | cathelicidin antimicrobial peptide |
Gene Aliases: |
CAP-18, CAP18, CRAMP, FALL-39, FALL39, HSD26, LL37 |
Location: |
Chr.3:48223347-48225491 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMP | NM_004345.4 | NP_004336.3 | ||
| BU074012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834682 | Chr.3:48096647 - 48269632 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map