Assay Details
Target Gene Details
Entrez Gene ID: | 3155 |
Gene Name: | 3-hydroxymethyl-3-methylglutaryl-CoA lyase |
Gene Aliases: |
HL |
Location: |
Chr.1:23801877-23825459 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGCL | NM_000191.2 | 9 | 1001 | NP_000182.2 |
| NM_001166059.1 | 7 | 788 | NP_001159531.1 | |
| AK122801.1 | 10 | 1554 | ||
| AK300733.1 | 7 | 757 | ||
| AK313869.1 | ||||
| BC010570.1 | 9 | 967 | AAH10570.1 | |
| BM981610.1 | 1 | 625 | ||
| BT009792.1 | AAP88794.1 | |||
| CA306929.1 | 1 | 622 | ||
| CR456884.1 | CAG33165.1 | |||
| L07033.1 | 9 | 971 | AAA92733.1 |
More Information
Set Membership: |
|

Genomic Map