Assay Details
Target Gene Details
Entrez Gene ID: | 57381 |
Gene Name: | ras homolog family member J |
Gene Aliases: |
ARHJ, RASL7B, TC10B, TCL |
Location: |
Chr.14:63204127-63293512 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOJ | NM_020663.4 | 1 | 76 | NP_065714.1 |
| XM_006720214.3 | 1 | 333 | XP_006720277.1 | |
| XM_011536993.2 | 1 | 333 | XP_011535295.1 | |
| AB209218.1 | BAD92455.1 | |||
| AF087862.1 | 1 | 52 | AAP97172.1 | |
| AK027278.1 | BAB55013.1 | |||
| AK027351.1 | BAB55055.1 | |||
| BC062575.1 | AAH62575.1 | |||
| BM555132.1 | 1 | 76 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv137n21 | Chr.14:62934068 - 63690399 on Build GRCh38 | Gain |
|
| nsv564893 | Chr.14:63060213 - 63329404 on Build GRCh38 | Gain |
|
| esv3567910 | Chr.14:62739985 - 63371766 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map