Assay Details
Target Gene Details
Entrez Gene ID: | 5934 |
Gene Name: | RB transcriptional corepressor like 2 |
Gene Aliases: |
P130, Rb2 |
Location: |
Chr.16:53434420-53491648 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBL2 | NM_001323608.1 | 6 | 990 | NP_001310537.1 |
| NM_001323609.1 | 6 | 990 | NP_001310538.1 | |
| NM_001323610.1 | 6 | 990 | NP_001310539.1 | |
| NM_001323611.1 | 6 | 739 | NP_001310540.1 | |
| NM_005611.3 | 6 | 971 | NP_005602.3 | |
| XM_011523253.2 | 4 | 364 | XP_011521555.1 | |
| XM_017023513.1 | 3 | 528 | XP_016879002.1 | |
| AK300047.1 | 6 | 709 | ||
| AK304283.1 | 3 | 438 | ||
| AK307881.1 | 6 | 948 | ||
| BC034490.1 | 6 | 961 | AAH34490.1 | |
| BX537767.1 | 4 | 430 | CAD97830.1 | |
| S67171.1 | 6 | 881 | AAB29227.1 | |
| X74594.1 | 6 | 682 | CAA52671.1 | |
| X76061.1 | 6 | 922 | CAA53661.1 |
More Information
Set Membership: |
|

Genomic Map