Assay Details
Target Gene Details
Entrez Gene ID: | 390038 |
Gene Name: | olfactory receptor family 51 subfamily D member 1 |
Gene Aliases: |
OR11-14, OR51D1Q |
Location: |
Chr.11:4639715-4640838 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51D1 | NM_001004751.2 | 1 | 901 | NP_001004751.1 |
| BC140717.1 | 1 | 901 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054705 | Chr.11:4560920 - 4666008 on Build GRCh38 | Loss |
|
| nsv1041633 | Chr.11:4618062 - 4671879 on Build GRCh38 | Gain |
|
| nsv1054847 | Chr.11:4621645 - 5179426 on Build GRCh38 | Gain |
|
| dgv1586n54 | Chr.11:4566710 - 4668512 on Build GRCh38 | Loss |
|
| nsv832055 | Chr.11:4529664 - 4715110 on Build GRCh38 | Loss |
|
| dgv1025n100 | Chr.11:4637957 - 4650235 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79020081] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map