Assay Details
Target Gene Details
Entrez Gene ID: | 7004 |
Gene Name: | TEA domain transcription factor 4 |
Gene Aliases: |
EFTR-2, RTEF1, TCF13L1, TEF-3, TEF3, TEFR-1, hRTEF-1B |
Location: |
Chr.12:2959312-3040676 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEAD4 | NM_003213.3 | 3 | 314 | NP_003204.2 |
| NM_201441.2 | 3 | 314 | NP_958849.1 | |
| NM_201443.2 | NP_958851.1 | |||
| AK222947.1 | 3 | 201 | BAD96667.1 | |
| AY101179.1 | 3 | 296 | AAM89497.1 | |
| BC015497.1 | 3 | 212 | ||
| BX640921.1 | CAE45959.1 | |||
| DA210894.1 | 3 | 314 | ||
| DA886817.1 | ||||
| U63824.1 | 3 | 185 | AAC50763.1 | |
| X94438.1 | CAA64212.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628332 | Chr.12:2965888 - 3003559 on Build GRCh38 | Gain |
|
| nsv1036000 | Chr.12:2964853 - 3039074 on Build GRCh38 | Gain |
|
| esv3628329 | Chr.12:2941695 - 3004851 on Build GRCh38 | Gain |
|
| nsv832311 | Chr.12:2810344 - 2996539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map