Assay Details
Target Gene Details
Entrez Gene ID: | 829 |
Gene Name: | capping actin protein of muscle Z-line alpha subunit 1 |
Gene Aliases: |
CAPPA1, CAPZ, CAZ1 |
Location: |
Chr.1:112619453-112671619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAPZA1 | NM_006135.2 | 1 | 148 | NP_006126.1 |
| BX648738.1 | 1 | 440 | ||
| DB233456.1 | 1 | 148 |
Target Gene Details
Entrez Gene ID: | 54879 |
Gene Name: | suppression of tumorigenicity 7 like |
Gene Aliases: |
FAM4B, ST7R, STLR |
Location: |
Chr.1:112517444-112620838 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST7L | XM_011541630.2 | XP_011539932.1 | ||
| XM_011541631.2 | XP_011539933.1 | |||
| BC037830.1 | AAH37830.1 | |||
| BX537988.1 | CAD97948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145904439] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map