Assay Details
Target Gene Details
Entrez Gene ID: | 4810 |
Gene Name: | NHS actin remodeling regulator |
Gene Aliases: |
CTRCT40, CXN, SCML1 |
Location: |
Chr.X:17375420-17735994 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NHS | NM_001291867.1 | 1 | 163 | NP_001278796.1 |
| NM_198270.3 | 1 | 163 | NP_938011.1 | |
| AK302437.1 | 1 | 349 | ||
| AY436752.1 | 1 | 163 | AAR03104.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1145285 | Chr.X:17370377 - 17376377 on Build GRCh38 | Deletion |
|
| dgv503n21 | Chr.X:17329494 - 17542854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map