Assay Details
Target Gene Details
Entrez Gene ID: | 79847 |
Gene Name: | major facilitator superfamily domain containing 13A |
Gene Aliases: |
C10orf77, TMEM180, bA18I14.8 |
Location: |
Chr.10:102460896-102480118 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD13A | NM_024789.3 | NP_079065.2 | ||
| XM_006717973.3 | XP_006718036.1 | |||
| XM_006717974.3 | XP_006718037.1 | |||
| XM_011540160.2 | XP_011538462.1 | |||
| XM_011540163.2 | XP_011538465.1 | |||
| XM_011540167.2 | XP_011538469.1 | |||
| XM_011540169.2 | XP_011538471.1 | |||
| XM_017016654.1 | XP_016872143.1 | |||
| XM_017016655.1 | XP_016872144.1 | |||
| XM_017016656.1 | XP_016872145.1 | |||
| XM_017016657.1 | XP_016872146.1 | |||
| XM_017016658.1 | XP_016872147.1 | |||
| XM_017016659.1 | XP_016872148.1 | |||
| AK026182.1 | BAB15385.1 | |||
| AK296583.1 | ||||
| AK301612.1 | ||||
| BC057787.1 | AAH57787.2 | |||
| BC067530.1 | ||||
| BC067531.1 | ||||
| BC113589.1 |

Genomic Map