Assay Details
Target Gene Details
Entrez Gene ID: | 11267 |
Gene Name: | SNF8, ESCRT-II complex subunit |
Gene Aliases: |
Dot3, EAP30, VPS22 |
Location: |
Chr.17:48930096-48944842 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNF8 | NM_001317192.1 | NP_001304121.1 | ||
| NM_001317193.1 | NP_001304122.1 | |||
| NM_001317194.1 | NP_001304123.1 | |||
| NM_007241.3 | NP_009172.2 | |||
| NR_133679.1 | ||||
| XM_017024075.1 | XP_016879564.1 | |||
| AF156102.1 | AAD46560.1 | |||
| BC008976.1 | AAH08976.1 | |||
| BC038830.1 | AAH38830.1 | |||
| BE394098.1 | ||||
| BE818533.1 | ||||
| BG248541.1 | ||||
| BP342490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2073 | Chr.17:48928271 - 48962703 on Build GRCh38 | Insertion |
|
| nsv833472 | Chr.17:48895840 - 49078381 on Build GRCh38 | Gain |
|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map