Assay Details
Target Gene Details
Entrez Gene ID: | 9685 |
Gene Name: | clathrin interactor 1 |
Gene Aliases: |
CLINT, ENTH, EPN4, EPNR |
Location: |
Chr.5:157785743-157859175 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLINT1 | NM_001195555.1 | 12 | 2382 | NP_001182484.1 |
| NM_001195556.1 | 12 | 2359 | NP_001182485.1 | |
| NM_014666.3 | 12 | 2328 | NP_055481.1 | |
| XM_011534701.2 | 12 | 2393 | XP_011533003.1 | |
| XM_017010087.1 | 12 | 2339 | XP_016865576.1 | |
| AK092765.1 | 12 | 2329 | BAC03971.1 | |
| AK128594.1 | 2 | 2609 | ||
| AL833548.1 | 2 | 1791 | ||
| BC013091.1 | 5 | 1193 | AAH13091.1 | |
| D79993.1 | 12 | 2224 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023774 | Chr.5:157782180 - 157852922 on Build GRCh38 | Loss |
|
| nsv521000 | Chr.5:157786489 - 157820856 on Build GRCh38 | Loss |
|
| nsv509097 | Chr.5:157767224 - 157800513 on Build GRCh38 | Insertion |
|
| esv2759389 | Chr.5:157570600 - 157806036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map