Assay Details
Target Gene Details
Entrez Gene ID: | 200035 |
Gene Name: | nudix hydrolase 17 |
Gene Aliases: |
- |
Location: |
Chr.1:145844985-145848954 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT17 | NM_001012758.2 | NP_001012776.1 | ||
| XM_011509260.2 | XP_011507562.1 | |||
| XM_011509262.1 | XP_011507564.1 | |||
| AK091752.1 | ||||
| AK298083.1 | ||||
| BG820863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| esv3587420 | Chr.1:145824369 - 145905171 on Build GRCh38 | Gain |
|
| nsv831381 | Chr.1:145784568 - 145960930 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map