Assay Details
Target Gene Details
Entrez Gene ID: | 6096 |
Gene Name: | RAR related orphan receptor B |
Gene Aliases: |
NR1F2, ROR-BETA, RZR-BETA, RZRB, bA133M9.1 |
Location: |
Chr.9:74497336-74687201 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORB | NM_006914.3 | 1 | 542 | NP_008845.2 |
| AK313431.1 | 1 | 206 | ||
| AL691650.1 | 1 | 543 | ||
| BC051830.1 | 1 | 530 | AAH51830.1 | |
| BC093772.1 | AAH93772.1 | |||
| BC093774.1 | AAH93774.1 | |||
| Y08639.1 | 1 | 498 | CAA69929.1 |
Target Gene Details
Entrez Gene ID: | 103752585 |
Gene Name: | RORB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:74473435-74498553 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RORB-AS1 | NR_125791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831622 | Chr.9:74434084 - 74532100 on Build GRCh38 | Gain |
|
| nsv614658 | Chr.9:74454955 - 74539535 on Build GRCh38 | Gain |
|
| dgv487n21 | Chr.9:74485743 - 74549090 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map