Assay Details
Target Gene Details
Entrez Gene ID: | 55584 |
Gene Name: | cholinergic receptor nicotinic alpha 9 subunit |
Gene Aliases: |
HSA243342, NACHRA9 |
Location: |
Chr.4:40335329-40355217 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNA9 | NM_017581.3 | NP_060051.2 | ||
| AF227732.1 | AAF61920.1 | |||
| AJ243342.1 | CAB65091.1 | |||
| BC113549.1 | ||||
| BC113575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569393 | Chr.4:40346277 - 40354016 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map