Assay Details
Target Gene Details
Entrez Gene ID: | 8218 |
Gene Name: | clathrin heavy chain like 1 |
Gene Aliases: |
CHC22, CLH22, CLTCL, CLTD |
Location: |
Chr.22:19179473-19291716 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLTCL1 | NM_001835.3 | 32 | 4965 | NP_001826.3 |
| NM_007098.3 | 33 | 5136 | NP_009029.3 | |
| XM_011530401.2 | 23 | 3370 | XP_011528703.1 | |
| XM_017028953.1 | 33 | 5105 | XP_016884442.1 | |
| XM_017028954.1 | 32 | 5015 | XP_016884443.1 | |
| XM_017028955.1 | 32 | 4913 | XP_016884444.1 | |
| XM_017028956.1 | 31 | 4847 | XP_016884445.1 | |
| AK125929.1 | 3 | 4259 | ||
| AK294147.1 | 10 | 1338 | ||
| AK295264.1 | 12 | 1618 | ||
| AK302506.1 | 32 | 4934 | ||
| L77568.1 | 19 | 2648 | ||
| U41763.1 | 33 | 5158 | AAC50494.1 | |
| U60802.1 | 33 | 5107 | AAB40908.1 | |
| X95486.1 | 33 | 5094 | CAA64752.1 | |
| X95487.1 | 5 | 542 | CAA64753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588213 | Chr.22:18971102 - 19298293 on Build GRCh38 | Gain |
|
| nsv834122 | Chr.22:19098491 - 19280189 on Build GRCh38 | Loss |
|
| esv3575418 | Chr.22:18161776 - 19963846 on Build GRCh38 | Gain |
|
| nsv588216 | Chr.22:19086099 - 19839117 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map