Assay Details
Target Gene Details
Entrez Gene ID: | 6820 |
Gene Name: | sulfotransferase family 2B member 1 |
Gene Aliases: |
HSST2 |
Location: |
Chr.19:48552172-48599427 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SULT2B1 | NM_177973.1 | NP_814444.1 | ||
| BC034694.1 | AAH34694.1 | |||
| U92315.1 | AAC78499.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3617n100 | Chr.19:48532114 - 48643732 on Build GRCh38 | Gain |
|
| nsv1057580 | Chr.19:48512123 - 48608737 on Build GRCh38 | Gain |
|
| nsv817853 | Chr.19:48510834 - 48574410 on Build GRCh38 | Loss |
|
| nsv1058767 | Chr.19:48519557 - 48562402 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map