Assay Details
Target Gene Details
Entrez Gene ID: | 11094 |
Gene Name: | calcium channel flower domain containing 1 |
Gene Aliases: |
C9orf7, D9S2135, FLOWER |
Location: |
Chr.9:133459965-133470787 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACFD1 | NM_001135775.2 | 4 | 773 | NP_001129247.1 |
| NM_001242369.1 | 6 | 1024 | NP_001229298.1 | |
| NM_001242370.1 | 5 | 898 | NP_001229299.1 | |
| NM_017586.3 | 5 | 899 | NP_060056.1 | |
| XM_011518181.2 | 6 | 2581 | XP_011516483.1 | |
| XM_011518182.1 | 6 | 1077 | XP_011516484.1 | |
| XM_011518184.1 | 5 | 1640 | XP_011516486.1 | |
| XM_011518185.2 | 5 | 2456 | XP_011516487.1 | |
| XM_011518186.1 | 5 | 952 | XP_011516488.1 | |
| AJ011373.1 | 5 | 855 | CAB66156.1 | |
| AK074852.1 | 5 | 877 | BAC11245.1 | |
| AK075416.1 | 4 | 771 | BAC11606.1 | |
| AK075548.1 | 5 | 867 | BAC11691.1 | |
| AK296343.1 | ||||
| AK298820.1 | 5 | 830 | ||
| AK304524.1 | 6 | 857 | ||
| AK311325.1 | 5 | 955 | ||
| AY358452.1 | 5 | 847 | AAQ88817.1 | |
| BC030558.1 | 5 | 865 | AAH30558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map