Assay Details
Target Gene Details
Entrez Gene ID: | 2662 |
Gene Name: | growth differentiation factor 10 |
Gene Aliases: |
BMP-3b, BMP3B |
Location: |
Chr.10:47300224-47313577 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF10 | NM_004962.3 | NP_004953.1 | ||
| AK315322.1 | ||||
| BC028237.1 | AAH28237.1 | |||
| D49492.1 | BAA08452.1 | |||
| L42113.1 | AAL77527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831861 | Chr.10:47306097 - 47471541 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map