Assay Details
Target Gene Details
Entrez Gene ID: | 374654 |
Gene Name: | kinesin family member 7 |
Gene Aliases: |
ACLS, AGBK, HLS2, JBTS12, UNQ340 |
Location: |
Chr.15:89627970-89663086 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF7 | NM_198525.2 | 17 | 3558 | NP_940927.2 |
| XM_011521531.2 | 17 | 3668 | XP_011519833.1 | |
| AY358384.1 | 13 | 2127 | AAQ88750.1 | |
| BC040878.1 | 5 | 857 | AAH40878.1 | |
| BC104044.1 | 13 | 2100 | AAI04045.1 | |
| BC112271.1 | 12 | 1991 | AAI12272.1 | |
| BC112273.1 | 12 | 1991 | AAI12274.1 | |
| BC143461.1 | 12 | 1987 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1659 | Chr.15:89610536 - 89655632 on Build GRCh38 | Deletion |
|
| esv3892721 | Chr.15:89530925 - 89673865 on Build GRCh38 | Gain |
|
| esv34025 | Chr.15:89412131 - 89630626 on Build GRCh38 | Loss |
|
| nsv521260 | Chr.15:89627006 - 89631197 on Build GRCh38 | Loss |
|
| nsv1160317 | Chr.15:89614949 - 89683716 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map