Assay Details
Target Gene Details
Entrez Gene ID: | 2916 |
Gene Name: | glutamate metabotropic receptor 6 |
Gene Aliases: |
CSNB1B, GPRC1F, MGLUR6, mGlu6 |
Location: |
Chr.5:178978327-178995123 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM6 | NM_000843.3 | 10 | 2844 | NP_000834.2 |
| BX647168.1 | 2 | 1388 | CAI46028.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600489 | Chr.5:178941095 - 179003374 on Build GRCh38 | Loss |
|
| nsv950734 | Chr.5:178981200 - 178999999 on Build GRCh38 | Deletion |
|
| nsv600490 | Chr.5:178978250 - 179003374 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map