Assay Details
Target Gene Details
Entrez Gene ID: | 10068 |
Gene Name: | interleukin 18 binding protein |
Gene Aliases: |
IL18BPa |
Location: |
Chr.11:71997944-72002919 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL18BP | NM_001039659.1 | NP_001034748.1 | ||
| NM_001039660.1 | NP_001034749.1 | |||
| NM_001145055.1 | NP_001138527.1 | |||
| NM_001145057.1 | NP_001138529.1 | |||
| NM_005699.3 | 1 | 906 | NP_005690.2 | |
| NM_173042.2 | 1 | 906 | NP_766630.2 | |
| NM_173044.2 | NP_766632.2 | |||
| XM_017017059.1 | 2 | 852 | XP_016872548.1 | |
| XM_017017060.1 | 2 | 819 | XP_016872549.1 | |
| XM_017017061.1 | 3 | 715 | XP_016872550.1 | |
| XM_017017062.1 | XP_016872551.1 | |||
| XM_017017063.1 | XP_016872552.1 | |||
| AB019504.1 | BAA76374.1 | |||
| AF110799.1 | AAD17190.1 | |||
| AF110800.1 | AAD17191.1 | |||
| AF110801.1 | 1 | 906 | AAD17192.1 | |
| AF122906.1 | AAD41051.1 | |||
| AF215907.2 | AAF31697.1 | |||
| AK098255.1 | 1 | 2001 | ||
| BC044215.1 | AAH44215.1 | |||
| BM819966.1 | ||||
| BX357194.2 | ||||
| CB154498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428262 | Chr.11:71652593 - 72093009 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map