Assay Details
Target Gene Details
Entrez Gene ID: | 27095 |
Gene Name: | trafficking protein particle complex 3 |
Gene Aliases: |
BET3 |
Location: |
Chr.1:36136569-36156053 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC3 | NM_001270894.1 | 5 | 770 | NP_001257823.1 |
| NM_001270895.1 | 5 | 739 | NP_001257824.1 | |
| NM_001270896.1 | 5 | 991 | NP_001257825.1 | |
| NM_001270897.1 | 3 | 548 | NP_001257826.1 | |
| NM_014408.4 | 5 | 746 | NP_055223.1 | |
| NR_073097.1 | 3 | 793 | ||
| NR_073098.1 | 2 | 365 | ||
| AA446176.1 | 2 | 319 | ||
| AF041432.1 | 5 | 701 | AAB96936.1 | |
| AJ224335.1 | 5 | 646 | CAA11902.1 | |
| AK094755.1 | 3 | 1873 | ||
| AY007139.1 | 5 | 680 | AAG02000.1 | |
| BC007662.2 | 5 | 698 | AAH07662.1 | |
| BG704657.1 | ||||
| BI762992.1 | ||||
| BM451581.1 | 5 | 739 | ||
| CD251784.1 | 5 | 788 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10106 | Chr.1:36134826 - 36151706 on Build GRCh38 | Gain |
|
| nsv545993 | Chr.1:36126011 - 36187018 on Build GRCh38 | Loss |
|
| nsv834691 | Chr.1:36059137 - 36238540 on Build GRCh38 | Loss |
|
| nsv545992 | Chr.1:36091266 - 36161590 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map