Assay Details
Target Gene Details
Entrez Gene ID: | 414899 |
Gene Name: | BH3-like motif containing, cell death inducer |
Gene Aliases: |
BRCC2 |
Location: |
Chr.11:122115354-122116215 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLID | NM_001001786.2 | 1 | 637 | NP_001001786.2 |
| AF303179.1 | 1 | 637 | AAN31127.1 | |
| BC130361.1 | ||||
| BC130363.1 |
Target Gene Details
Entrez Gene ID: | 399959 |
Gene Name: | mir-100-let-7a-2 cluster host gene |
Gene Aliases: |
AGD1, linc-NeD125, lncRNA-N2 |
Location: |
Chr.11:122089103-122203062 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR100HG | NR_024430.2 | |||
| NR_137179.1 | ||||
| NR_137180.1 | ||||
| NR_137181.1 | ||||
| NR_137182.1 | ||||
| NR_137183.1 | ||||
| NR_137184.1 | ||||
| NR_137185.1 | ||||
| NR_137186.1 | ||||
| NR_137187.1 | ||||
| NR_137188.1 | ||||
| NR_137192.1 | ||||
| NR_137193.1 | ||||
| NR_137194.1 | ||||
| AB485963.1 | ||||
| AK091713.1 | ||||
| AK311309.1 | ||||
| DA027094.1 | ||||
| DA153922.1 | ||||
| DA349788.1 | ||||
| DA474693.1 | ||||
| DA597554.1 | ||||
| DA649890.1 | ||||
| DA650430.1 | ||||
| DA659486.1 | ||||
| DA788103.1 | ||||
| DA808613.1 | ||||
| DA861327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv17n68 | Chr.11:122091983 - 122336712 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map