Assay Details
Target Gene Details
Entrez Gene ID: | 51320 |
Gene Name: | mex-3 RNA binding family member C |
Gene Aliases: |
BM-013, MEX-3C, RKHD2, RNF194 |
Location: |
Chr.18:51174550-51197681 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEX3C | NM_016626.4 | 2 | 3291 | NP_057710.3 |
| AF208855.1 | 1 | 1410 | AAF64269.1 | |
| AK096190.1 | 2 | 2532 | ||
| AL833519.1 | 1 | 2151 | ||
| BC041122.1 | 1 | 1582 | AAH41122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833652 | Chr.18:51146567 - 51296721 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map