Assay Details
Target Gene Details
Entrez Gene ID: | 26589 |
Gene Name: | mitochondrial ribosomal protein L46 |
Gene Aliases: |
C15orf4, LIECG2, P2ECSL |
Location: |
Chr.15:88459478-88467402 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL46 | NM_022163.3 | 2 | 392 | NP_071446.2 |
| AF205435.1 | 2 | 370 | AAG33698.1 | |
| AF210056.1 | 2 | 378 | AAG43507.1 | |
| AK074053.1 | 1 | 113 | BAB84879.1 | |
| AK315434.1 | 2 | 377 | ||
| BC017883.1 | 2 | 378 | AAH17883.1 | |
| CN309742.1 | 2 | 433 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833085 | Chr.15:88365076 - 88521662 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map