Assay Details
Target Gene Details
Entrez Gene ID: | 64766 |
Gene Name: | S100P binding protein |
Gene Aliases: |
S100PBPR |
Location: |
Chr.1:32816767-32858879 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100PBP | NM_001256121.1 | NP_001243050.1 | ||
| NM_022753.3 | NP_073590.2 | |||
| NR_135106.1 | ||||
| NR_135107.1 | ||||
| XM_011541961.2 | XP_011540263.1 | |||
| XM_011541962.2 | XP_011540264.1 | |||
| XM_011541963.1 | XP_011540265.1 | |||
| XM_017002098.1 | XP_016857587.1 | |||
| AK022965.1 | BAB14335.1 | |||
| AK090711.1 | ||||
| BC015175.1 | AAH15175.1 | |||
| BX647916.1 | ||||
| KU178750.1 | ||||
| KU178751.1 |
More Information
Set Membership: |
|

Genomic Map