Assay Details
Target Gene Details
Entrez Gene ID: | 284119 |
Gene Name: | polymerase I and transcript release factor |
Gene Aliases: |
CAVIN, CAVIN1, CGL4, FKSG13, cavin-1 |
Location: |
Chr.17:42402449-42423320 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRF | NM_012232.5 | 2 | 1658 | NP_036364.2 |
| AF312393.1 | 2 | 1855 | AAG27093.1 | |
| AK000715.1 | 2 | 1144 | ||
| AK092406.1 | ||||
| AK298072.1 | 3 | 1539 | ||
| AK298563.1 | 3 | 1513 | ||
| BC004295.1 | 1 | 868 | AAH04295.1 | |
| BC008849.1 | 2 | 941 | AAH08849.1 | |
| BC066123.1 | 2 | 1594 | AAH66123.1 | |
| BC073759.1 | 2 | 1480 | AAH73759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470586 | Chr.17:42362183 - 42459270 on Build GRCh38 | Gain |
|
| nsv833449 | Chr.17:42274854 - 42421081 on Build GRCh38 | Loss |
|
| nsv575051 | Chr.17:42317580 - 42498404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map