Assay Details
Target Gene Details
Entrez Gene ID: | 144165 |
Gene Name: | prickle planar cell polarity protein 1 |
Gene Aliases: |
EPM1B, RILP |
Location: |
Chr.12:42458338-42589770 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE1 | NM_001144881.1 | NP_001138353.1 | ||
| NM_001144882.1 | NP_001138354.1 | |||
| NM_001144883.1 | NP_001138355.1 | |||
| NM_153026.2 | NP_694571.2 | |||
| XM_011537946.1 | XP_011536248.1 | |||
| XM_011537947.2 | XP_011536249.1 | |||
| XM_017018838.1 | XP_016874327.1 | |||
| XM_017018839.1 | XP_016874328.1 | |||
| XM_017018840.1 | XP_016874329.1 | |||
| AF399844.1 | AAQ03035.1 | |||
| AK056499.1 | BAB71198.1 | |||
| AK122877.1 | ||||
| AK122881.1 | ||||
| BC114939.1 | ||||
| BC114940.1 | ||||
| CN390789.1 |
More Information
Set Membership: |
|

Genomic Map