Assay Details
Target Gene Details
Entrez Gene ID: | 8822 |
Gene Name: | fibroblast growth factor 17 |
Gene Aliases: |
FGF-13, FGF-17, HH20 |
Location: |
Chr.8:22039708-22048809 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF17 | NM_001304478.1 | NP_001291407.1 | ||
| NM_003867.3 | NP_003858.1 | |||
| XM_011544683.1 | XP_011542985.1 | |||
| XM_011544684.2 | XP_011542986.1 | |||
| XM_011544685.1 | XP_011542987.1 | |||
| AB009249.1 | BAA25429.1 | |||
| AY358869.1 | AAQ89228.1 | |||
| BC069475.1 | AAH69475.1 | |||
| BC105131.1 | AAI05132.1 | |||
| BC113489.1 | ||||
| BC143789.1 | ||||
| DR159037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528590 | Chr.8:22034428 - 22168131 on Build GRCh38 | Loss |
|
| nsv610764 | Chr.8:22031342 - 22094715 on Build GRCh38 | Loss |
|
| nsv951112 | Chr.8:22023690 - 22135287 on Build GRCh38 | Deletion |
|
| nsv831263 | Chr.8:22031088 - 22184413 on Build GRCh38 | Loss |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map