Assay Details
Target Gene Details
Entrez Gene ID: | 27440 |
Gene Name: | cat eye syndrome chromosome region, candidate 5 |
Gene Aliases: |
- |
Location: |
Chr.22:17137520-17165287 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR5 | NM_017829.5 | 8 | 1211 | NP_060299.4 |
| NM_033070.2 | 8 | 1286 | NP_149061.1 | |
| XM_005261236.3 | 8 | 1264 | XP_005261293.1 | |
| XM_011546126.2 | 7 | 1082 | XP_011544428.1 | |
| XM_011546127.1 | 8 | 1701 | XP_011544429.1 | |
| XM_017028781.1 | 8 | 1753 | XP_016884270.1 | |
| XM_017028782.1 | 9 | 2080 | XP_016884271.1 | |
| XM_017028783.1 | 9 | 2517 | XP_016884272.1 | |
| XM_017028784.1 | 9 | 2080 | XP_016884273.1 | |
| AF273270.1 | 8 | 1211 | AAK19151.1 | |
| AF273271.1 | 8 | 1271 | AAK19152.1 | |
| AK000461.1 | 8 | 1284 | BAA91180.1 | |
| AK001034.1 | 8 | 1264 | BAA91475.1 | |
| AK123646.1 | 3 | 1767 | ||
| AK315156.1 | ||||
| AL833450.1 | 3 | 3102 | ||
| BC042540.1 | 8 | 1270 | AAH42540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828924 | Chr.22:17116393 - 17174153 on Build GRCh38 | Gain |
|
| nsv1064641 | Chr.22:16757380 - 17233805 on Build GRCh38 | Gain |
|
| esv3647208 | Chr.22:17069325 - 17142761 on Build GRCh38 | Gain |
|
| nsv526946 | Chr.22:17137252 - 17153160 on Build GRCh38 | Loss |
|
| nsv509805 | Chr.22:17081417 - 17267685 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map