Assay Details
Target Gene Details
Entrez Gene ID: | 11010 |
Gene Name: | GLI pathogenesis related 1 |
Gene Aliases: |
CRISP7, GLIPR, RTVP1 |
Location: |
Chr.12:75480733-75501936 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIPR1 | NM_006851.2 | 1 | 88 | NP_006842.2 |
| XM_011537775.2 | 1 | 67 | XP_011536077.1 | |
| AF400440.1 | AAK92489.1 | |||
| AK304847.1 | 1 | 68 | ||
| AK313449.1 | ||||
| BC012510.1 | AAH12510.1 | |||
| CB123261.1 | 1 | 141 | ||
| EF667354.1 | ||||
| U16307.3 | AAA82731.3 | |||
| X91911.1 | 1 | 68 | CAA63005.1 |
More Information
Set Membership: |
|

Genomic Map