Assay Details
Target Gene Details
Entrez Gene ID: | 81030 |
Gene Name: | Z-DNA binding protein 1 |
Gene Aliases: |
C20orf183, DAI, DLM-1, DLM1 |
Location: |
Chr.20:57603846-57620576 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBP1 | NM_001160417.1 | 7 | 1200 | NP_001153889.1 |
| NM_001160418.1 | 6 | 978 | NP_001153890.1 | |
| NM_030776.2 | 7 | 1203 | NP_110403.2 | |
| NR_136660.1 | 6 | 978 | ||
| XM_011529057.2 | 7 | 1053 | XP_011527359.1 | |
| XM_011529058.2 | 8 | 1216 | XP_011527360.1 | |
| AJ300575.1 | 7 | 1100 | CAC18810.1 | |
| AK122761.1 | 6 | 981 | ||
| AK128405.1 | 6 | 3326 | ||
| AK290918.1 | 7 | 1053 | ||
| AK314062.1 | 7 | 1022 | ||
| AX775973.1 | 7 | 1100 | CAE11686.1 | |
| AX775975.1 | 7 | 1100 | CAE11687.1 | |
| BC131706.1 | 7 | 1018 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758802 | Chr.20:57595722 - 58113215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map