Assay Details
Target Gene Details
Entrez Gene ID: | 51371 |
Gene Name: | proteasome maturation protein |
Gene Aliases: |
C13orf12, HSPC014, PNAS-110, UMP1 |
Location: |
Chr.13:28659004-28678957 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POMP | NM_015932.5 | NP_057016.1 | ||
| AF077200.1 | AAD26995.1 | |||
| AF125097.1 | AAD39914.1 | |||
| AF262975.1 | AAK58521.1 | |||
| AF275807.1 | AAG23819.1 | |||
| AK312118.1 | ||||
| BC003390.1 | AAH03390.1 | |||
| BC014334.1 | AAH14334.1 | |||
| EF535527.1 | ||||
| GU045555.1 | ||||
| GU045556.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832571 | Chr.13:28675794 - 28845425 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map