Assay Details
Target Gene Details
Entrez Gene ID: | 11275 |
Gene Name: | kelch like family member 2 |
Gene Aliases: |
ABP-KELCH, MAV, MAYVEN |
Location: |
Chr.4:165207618-165323156 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL2 | NM_001161521.1 | 12 | 1521 | NP_001154993.1 |
| NM_001161522.1 | 11 | 1414 | NP_001154994.1 | |
| NM_007246.3 | 12 | 1644 | NP_009177.3 | |
| XM_011531572.2 | 13 | 1634 | XP_011529874.1 | |
| XM_011531575.2 | 11 | 1466 | XP_011529877.1 | |
| XM_011531576.2 | 10 | 1134 | XP_011529878.1 | |
| XM_017007674.1 | 13 | 1594 | XP_016863163.1 | |
| XM_017007676.1 | 11 | 1467 | XP_016863165.1 | |
| XM_017007677.1 | 10 | 1341 | XP_016863166.1 | |
| AB209727.1 | 8 | 988 | BAD92964.1 | |
| AF059569.1 | 12 | 1493 | AAC67502.1 | |
| AK025289.1 | 9 | 1110 | ||
| AK294103.1 | 12 | 1515 | ||
| AK294336.1 | 8 | 943 | ||
| AK308940.1 | 7 | 826 | ||
| AK315372.1 | 12 | 1489 | ||
| BC022503.1 | 12 | 1626 | AAH22503.1 | |
| BC036468.2 | 12 | 1585 | AAH36468.1 | |
| BX647776.1 | 11 | 1398 |
More Information
Set Membership: |
|

Genomic Map