Assay Details
Target Gene Details
Entrez Gene ID: | 7169 |
Gene Name: | tropomyosin 2 (beta) |
Gene Aliases: |
AMCD1, DA1, DA2B, HEL-S-273, NEM4, TMSB |
Location: |
Chr.9:35681993-35690056 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPM2 | NM_001301226.1 | NP_001288155.1 | ||
| NM_001301227.1 | NP_001288156.1 | |||
| NM_003289.3 | NP_003280.2 | |||
| NM_213674.1 | NP_998839.1 | |||
| XM_017015087.1 | XP_016870576.1 | |||
| XM_017015088.1 | XP_016870577.1 | |||
| XM_017015090.1 | XP_016870579.1 | |||
| XM_017015091.1 | XP_016870580.1 | |||
| XM_017015092.1 | XP_016870581.1 | |||
| XM_017015093.1 | XP_016870582.1 | |||
| AI360640.1 | ||||
| AJ572605.1 | ||||
| AK130045.1 | ||||
| AK223258.1 | BAD96978.1 | |||
| AK300249.1 | ||||
| AK304803.1 | ||||
| BC011776.2 | AAH11776.1 | |||
| BE294047.1 | ||||
| EU106177.1 | ||||
| F24979.1 | ||||
| FJ224304.1 | ||||
| M12125.1 | AAA36773.1 | |||
| M12126.1 | AAA61229.1 | |||
| M74817.1 | AAA61230.1 | |||
| M75165.1 | AAB59509.1 | |||
| X06825.1 | CAA29971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6528 | Chr.9:35666752 - 35700672 on Build GRCh38 | Insertion |
|
| nsv614166 | Chr.9:35662254 - 35705089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map