Assay Details
Target Gene Details
Entrez Gene ID: | 84270 |
Gene Name: | caspase recruitment domain family member 19 |
Gene Aliases: |
BinCARD, C9orf89 |
Location: |
Chr.9:93096123-93113295 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD19 | NM_001318010.1 | NP_001304939.1 | ||
| NM_001318011.1 | NP_001304940.1 | |||
| NM_032310.4 | NP_115686.3 | |||
| NR_134464.1 | 3 | 1719 | ||
| NR_134465.1 | 3 | 1708 | ||
| NR_134466.1 | ||||
| NR_134467.1 | ||||
| NR_134468.1 | ||||
| AA804861.1 | ||||
| AK057716.1 | 3 | 1614 | BAB71550.1 | |
| AK091611.1 | 3 | 1625 | ||
| AK097977.1 | 1 | 2011 | ||
| AK128259.1 | 1 | 2747 | ||
| BC004500.2 | AAH04500.1 | |||
| BM930683.1 | ||||
| BX354102.2 | ||||
| CD676329.1 | ||||
| DB550397.1 | ||||
| DN998463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526296 | Chr.9:92945910 - 93268110 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map