Assay Details
Target Gene Details
Entrez Gene ID: | 30827 |
Gene Name: | CXXC finger protein 1 |
Gene Aliases: |
2410002I16Rik, 5830420C16Rik, CFP1, CGBP, HsT2645, PCCX1, PHF18, SPP1, ZCGPC1, hCGBP |
Location: |
Chr.18:50282343-50288322 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXXC1 | NM_001101654.1 | NP_001095124.1 | ||
| NM_014593.3 | NP_055408.2 | |||
| XM_011525940.2 | XP_011524242.1 | |||
| XM_011525941.2 | XP_011524243.1 | |||
| XM_017025718.1 | XP_016881207.1 | |||
| XM_017025719.1 | XP_016881208.1 | |||
| AB031069.1 | BAA96307.1 | |||
| AB209883.1 | BAD93120.1 | |||
| AF149758.1 | AAF37799.1 | |||
| AK294645.1 | ||||
| AK314886.1 | ||||
| AL136862.1 | CAB66796.1 | |||
| BC014940.2 | AAH14940.1 | |||
| BC015733.1 | AAH15733.1 | |||
| BX647189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953541 | Chr.18:50272231 - 50289530 on Build GRCh38 | Deletion |
|
| nsv576889 | Chr.18:50227099 - 50287465 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map