Assay Details
Target Gene Details
Entrez Gene ID: | 23415 |
Gene Name: | potassium voltage-gated channel subfamily H member 4 |
Gene Aliases: |
BEC2, ELK1, Kv12.3 |
Location: |
Chr.17:42156892-42181278 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH4 | NM_012285.2 | NP_036417.1 | ||
| XM_017024400.1 | 16 | 3637 | XP_016879889.1 | |
| XM_017024401.1 | 15 | 3490 | XP_016879890.1 | |
| XM_017024402.1 | 16 | 3303 | XP_016879891.1 | |
| AB022698.1 | BAA83592.1 | |||
| AW084816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map