Assay Details
Target Gene Details
Entrez Gene ID: | 5609 |
Gene Name: | mitogen-activated protein kinase kinase 7 |
Gene Aliases: |
JNKK2, MAPKK7, MEK, MEK 7, MKK7, PRKMK7, SAPKK-4, SAPKK4 |
Location: |
Chr.19:7903780-7914483 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP2K7 | NM_001297555.1 | NP_001284484.1 | ||
| NM_001297556.1 | NP_001284485.1 | |||
| NM_145185.3 | NP_660186.1 | |||
| XM_006722800.2 | XP_006722863.1 | |||
| AF006689.1 | AAB97813.1 | |||
| AF013588.1 | AAC16272.1 | |||
| AF013589.1 | AAC16273.1 | |||
| AF014401.1 | AAB88048.1 | |||
| AF022805.1 | AAC26142.1 | |||
| AK300988.1 | ||||
| AK313899.1 | ||||
| BC038295.2 | AAH38295.1 | |||
| DQ445915.1 | ABE03013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953961 | Chr.19:7881316 - 7955515 on Build GRCh38 | Deletion |
|
| nsv833733 | Chr.19:7799229 - 7995254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map