Assay Details
Target Gene Details
Entrez Gene ID: | 165530 |
Gene Name: | C-type lectin domain family 4 member F |
Gene Aliases: |
CLECSF13, KCLR |
Location: |
Chr.2:70808643-70825235 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC4F | NM_001258027.1 | NP_001244956.1 | ||
| NM_001321308.1 | NP_001308237.1 | |||
| NM_173535.2 | NP_775806.2 | |||
| XM_011532635.2 | XP_011530937.1 | |||
| XM_011532637.2 | XP_011530939.1 | |||
| XM_011532638.2 | XP_011530940.1 | |||
| XM_011532639.2 | XP_011530941.1 | |||
| XM_011532640.2 | XP_011530942.1 | |||
| XM_011532641.2 | XP_011530943.1 | |||
| XM_011532642.2 | XP_011530944.1 | |||
| XM_011532643.1 | XP_011530945.1 | |||
| XM_017003519.1 | XP_016859008.1 | |||
| AK096429.1 | BAC04786.1 | |||
| AK301242.1 | ||||
| BC139723.1 | ||||
| BC144652.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762674 | Chr.2:70752618 - 70829950 on Build GRCh38 | Gain |
|
| nsv961756 | Chr.2:70813066 - 70829853 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map