Assay Details
Target Gene Details
Entrez Gene ID: | 3357 |
Gene Name: | 5-hydroxytryptamine receptor 2B |
Gene Aliases: |
5-HT(2B), 5-HT-2B, 5-HT2B |
Location: |
Chr.2:231108230-231125778 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR2B | NM_000867.4 | 4 | 1805 | NP_000858.3 |
| NM_001320758.1 | 4 | 1422 | NP_001307687.1 | |
| XM_005246520.3 | 3 | 1755 | XP_005246577.1 | |
| XM_006712482.2 | 4 | 2002 | XP_006712545.1 | |
| AK313741.1 | 4 | 1737 | ||
| AY136751.1 | 3 | 1300 | AAN01277.1 | |
| BC063123.1 | 4 | 1809 | AAH63123.1 | |
| X77307.1 | 3 | 1355 | CAA54513.1 | |
| Z36748.1 | 3 | 1346 | CAA85319.1 |
Target Gene Details
Entrez Gene ID: | 5707 |
Gene Name: | proteasome 26S subunit, non-ATPase 1 |
Gene Aliases: |
P112, Rpn2, S1 |
Location: |
Chr.2:231056864-231172827 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD1 | NM_001191037.1 | NP_001177966.1 | ||
| NM_002807.3 | NP_002798.2 | |||
| NR_034059.1 | ||||
| AK307415.1 | ||||
| AK307513.1 | ||||
| AK312528.1 | ||||
| BC001053.2 | AAH01053.1 | |||
| BC014013.2 | AAH14013.1 | |||
| BC020223.1 | ||||
| BC032445.1 | ||||
| BC039845.1 | AAH39845.1 | |||
| BC047897.1 | AAH47897.1 | |||
| BC064398.1 | AAH64398.1 | |||
| BC073833.1 | AAH73833.1 | |||
| BC094720.1 | AAH94720.1 | |||
| BC112344.1 | AAI12345.1 | |||
| BC114434.1 | AAI14435.1 | |||
| D44466.1 | BAA07918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524208 | Chr.2:231077778 - 231171079 on Build GRCh38 | Gain |
|
| nsv525474 | Chr.2:231062855 - 231171079 on Build GRCh38 | Loss |
|
| dgv4181n100 | Chr.2:231055301 - 231131114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map