Assay Details
Target Gene Details
Entrez Gene ID: | 83988 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
- |
Location: |
Chr.8:101686542-102124907 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCALD | NM_001040624.1 | 8 | 3130 | NP_001035714.1 |
| NM_001040625.1 | 7 | 3043 | NP_001035715.1 | |
| NM_001040626.1 | 7 | 3043 | NP_001035716.1 | |
| NM_001040627.1 | 6 | 2956 | NP_001035717.1 | |
| NM_001040628.1 | 8 | 3099 | NP_001035718.1 | |
| NM_001040629.1 | 7 | 3012 | NP_001035719.1 | |
| NM_001040630.1 | 6 | 3196 | NP_001035720.1 | |
| NM_032041.2 | 4 | 3105 | NP_114430.2 | |
| XM_011517332.2 | 4 | 2894 | XP_011515634.1 | |
| XM_011517333.2 | 8 | 3041 | XP_011515635.1 | |
| XM_011517334.2 | 5 | 2925 | XP_011515636.1 | |
| XM_011517335.2 | 6 | 3017 | XP_011515637.1 | |
| XM_017013901.1 | 7 | 3233 | XP_016869390.1 | |
| XM_017013902.1 | 5 | 2904 | XP_016869391.1 | |
| AB209015.1 | 5 | 5921 | BAD92252.1 | |
| AF052142.1 | 1 | 1121 | ||
| AF251061.1 | 6 | 2884 | AAK34951.1 | |
| AK055073.1 | 5 | 2723 | ||
| AK096515.1 | 8 | 3055 | ||
| AL136612.1 | 4 | 2856 | CAB66547.1 | |
| BC036098.1 | 6 | 2883 | AAH36098.1 | |
| BM681731.1 | 1 | 631 |
More Information
Set Membership: |
|

Genomic Map