Assay Details
Target Gene Details
Entrez Gene ID: | 25957 |
Gene Name: | PNN interacting serine and arginine rich protein |
Gene Aliases: |
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2 |
Location: |
Chr.6:99398051-99425345 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNISR | NM_001322405.1 | 12 | 2133 | NP_001309334.1 |
| NM_001322406.1 | 13 | 2249 | NP_001309335.1 | |
| NM_001322408.1 | 11 | 2053 | NP_001309337.1 | |
| NM_001322410.1 | 12 | 2137 | NP_001309339.1 | |
| NM_001322412.1 | 12 | 2133 | NP_001309341.1 | |
| NM_001322413.1 | 11 | 2057 | NP_001309342.1 | |
| NM_001322414.1 | 11 | 2083 | NP_001309343.1 | |
| NM_001322415.1 | 10 | 2003 | NP_001309344.1 | |
| NM_001322416.1 | NP_001309345.1 | |||
| NM_015491.2 | 11 | 2057 | NP_056306.1 | |
| NM_032870.3 | 12 | 2137 | NP_116259.2 | |
| NR_136326.1 | 10 | 2777 | ||
| XM_005266912.3 | 11 | 2016 | XP_005266969.2 | |
| XM_017010710.1 | 11 | 2080 | XP_016866199.1 | |
| XM_017010711.1 | 10 | 1966 | XP_016866200.1 | |
| XM_017010712.1 | 12 | 3488 | XP_016866201.1 | |
| XM_017010713.1 | 12 | 3484 | XP_016866202.1 | |
| AA626029.1 | ||||
| AF314184.1 | 10 | 1927 | AAL76163.1 | |
| AF314185.1 | 1 | 329 | AAL76164.1 | |
| AJ420559.1 | 1 | 1376 | ||
| AL080186.1 | 1 | 378 | CAB45767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890914 | Chr.6:99393043 - 99409897 on Build GRCh38 | Loss |
|
| nsv527517 | Chr.6:99394180 - 99416102 on Build GRCh38 | Loss |
|
| nsv1029536 | Chr.6:98422726 - 99818247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map